Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF2A Monoclonal Antibody (3A7A8)
GREENER_CHOICE

Catalog No. PIMA531725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531725 Supplier Invitrogen™ Supplier No. MA531725
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31725 has been tested in indirect ELISA.

EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF2A
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3A7A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene EIF2A
Gene Accession No. Q9BY44
Gene Alias 0910001O23Rik; 2410026C18Rik; 35kDa; 65 kDa eukaryotic translation initiation factor 2A; CDA02; D030048D22; D3Ertd194e; EIF2; EIF-2; EIF2A; eIF-2A; eIF2alpha; eIF-2alpha; eIF-2-alpha; EIF2S1; EIF2S1 protein; eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed; eukaryotic translation initiation factor subunit 1-like protein; initiation factor 2 alpha; MST089; MSTP004; MSTP089
Gene Symbols EIF2A
Host Species Mouse
Immunogen Purified recombinant fragment of human EIF2A (AA: 448-576) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.