Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2A Mouse anti-Human, Monkey, Mouse, Rat, Clone:3A7B11, Abnova™

Catalog No. 89334472 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-472 Supplier Abnova Corporation Supplier No. MAB17787
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human EIF2A.

EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes (Zoll et al., 2002 [PubMed 12133843]).[supplied by OMIM]

Specifications

Antigen EIF2A
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A7B11
Conjugate Unconjugated
Description eukaryotic translation initiation factor 2A, 65kDa
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene EIF2A
Gene Alias CDA02, EIF-2A, MST089, MSTP004, MSTP089
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 448-576 of human EIF2A from E. coli.
Quantity 100 μg
Gene ID (Entrez) 83939
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.