Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF2A, p Ser51 Antibody (SZ01-06), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267353
Description
eIF2A Monoclonal specifically detects eIF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
eIF2A (p Ser51) | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:500 | |
EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD ) | |
Rabbit | |
Protein A purified | |
Chromatin Research, DNA Repair, Lipid and Metabolism, Phospho Specific, Translation Control, Unfolded Protein Response | |
83939 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
SZ01-06 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
EIF2A | |
Synthetic phospho-peptide corresponding to residues surrounding Ser51 of Human eIF2A. (SwissProt: P05198 Human; SwissProt: Q6ZWX6 Mouse; SwissProt: P68101 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction