Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF2S1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527366

Catalog No. PIPA527366


Only null left
Add to Cart

Description

Description

Recommended positive controls: A549, H1299, HCT-116, rat neuron, HeLa, N2a. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (92%), Chicken (97%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EIF2S1
Polyclonal
Unconjugated
EIF2S1
0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01
Rabbit
Antigen affinity chromatography
RUO
13665, 1965, 54318
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.63 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P05198, P68101, Q6ZWX6
EIF2S1
Recombinant protein encompassing a sequence within the center region of human eIF2 alpha. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.