Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EIF2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541916
Description
Peptide sequence: DKSPDLAPTP APQSTPRNTV SQSISGDPEI DKKIKNLKKK LKAIEQLKEQ Sequence homology: Cow: 86%; Dog: 100%; Goat: 83%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
Specifications
| EIF2A | |
| Polyclonal | |
| Unconjugated | |
| EIF2A | |
| 0910001O23Rik; 2410026C18Rik; 35kDa; 65 kDa eukaryotic translation initiation factor 2A; CDA02; D030048D22; D3Ertd194e; EIF2; EIF-2; EIF2A; eIF-2A; eIF2alpha; eIF-2alpha; eIF-2-alpha; EIF2S1; EIF2S1 protein; eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed; eukaryotic translation initiation factor subunit 1-like protein; initiation factor 2 alpha; MST089; MSTP004; MSTP089 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 83939 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BY44 | |
| EIF2A | |
| Synthetic peptide directed towards the C-terminal of human EIF2A (aa 504-553). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction