Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2A Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991589
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1589 100 μg
50-199-1590 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1589 Supplier Rockland Immunochemicals Supplier No. 600101ML7
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human EIF2A. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 86.7% homology with the immunizing sequence to mouse.

Specifications

Antigen EIF2A
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene EIF2A
Gene Accession No. Q8BJW6, Q9BY44
Gene Alias 65 kDa eukaryotic translation initiation factor 2A; CDA02; D030048D22; D3Ertd194e; Eif2a; eIF-2A; Eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed; MST089; MSTP004; MSTP089
Gene Symbols EIF2A
Host Species Goat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229317, 83939
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.