Learn More
Promega EIF2AK2 Kinase Enzyme System, 10microg
Catalog No. PRV5328 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV5328 10 μg
1 options
Catalog No. PRV5328 Supplier Promega Supplier No. V5328
Recombinant human EIF2AK2 (252-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EIF2AK2 (also known as double-stranded RNA-activated protein kinase) is shown to be involved in HIV/gp120-associated neurodegeneration.
  • Easily Screen and Profile EIF2AK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C