Learn More
Promega EIF2AK2 Kinase Enzyme System, 1mg
Catalog No. PRV6585 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6585 1 mg
1 options
Catalog No. PRV6585 Supplier Promega Supplier No. V6585
Add to Cart
Add to Cart
Request a Quote
Recombinant human EIF2AK2 (252-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EIF2AK2 (also known as double-stranded RNA-activated protein kinase) is shown to be involved in HIV/gp120-associated neurodegeneration.
  • Easily Screen and Profile EIF2AK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg