Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF2AK3 Mouse anti-Human, Clone: 3C3, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM51385M

Description
PERK, a member of the GCN2 subfamily of Ser/Thr protein kinases, phosphorylates the alpha subunit of eukaryotic translation-initiation factor 2 (EIF2), leading to its inactivation and thus to a rapid reduction of translational initiation and repression of global protein synthesis. It likely serves as a critical effector of unfolded protein response (UPR)-induced G1 growth arrest due to the loss of cyclin D1 Perturbation in protein folding in the endoplasmic reticulum (ER) promotes reversible dissociation from HSPA5/BIP and oligomerization, resulting in transautophosphorylation and kinase activity induction Expression of this Type I membrane protein is ubiquitous, with highest levels seen in secretory tissues. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.Specifications
EIF2AK3 | |
Monoclonal | |
1 μg/mL | |
TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH | |
Q9NZJ5 | |
EIF2AK3 | |
This EIF2AK3 antibody is generated from a mouse immunized with a recombinant protein conjugated synthetic peptide between 530-850 amino acids from human EIF2AK3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3C3 | |
Unconjugated | |
EIF2AK3 | |
2.7.11.1; AI427929; Eif2ak3; eukaryotic translation initiation factor 2 alpha kinase 3; Eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; PERK; PRKR-like endoplasmic reticulum kinase; WRS | |
Mouse | |
Protein G | |
RUO | |
9451 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction