Learn More
eIF2alpha/EIF2S1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300721AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.Specifications
| eIF2alpha/EIF2S1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P05198, Q6ZWX6 | |
| Eif2s1 | |
| Between 275 and C-term | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| EIF2S1 | |
| EIF-2, EIF-2A, EIF-2alpha, EIF2, EIF2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13665, 1965 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.