Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF2b alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571697
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571697 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571697 Supplier Invitrogen™ Supplier No. PA571697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF2b alpha Polyclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes one of five subunits of eukaryotic translation initiation factor 2B (EIF2B), a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2b alpha
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EIF2B1
Gene Accession No. Q14232
Gene Alias 26kDa; D5Ertd406e; eIF-2a; EIF2B; eIF-2B GDP-GTP exchange factor subunit alpha; Eif2b1; EIF2BA; eIF-2Balpha; eukaryotic translation initiation factor 2B subunit alpha; eukaryotic translation initiation factor 2B, subunit 1 (alpha); eukaryotic translation initiation factor 2B, subunit 1 (alpha, 26kDa); eukaryotic translation initiation factor 2B, subunit 1 alpha; eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa; GTP-exchange protein; translation initiation factor eIF-2B subunit alpha
Gene Symbols EIF2B1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 136-163 amino acids from the Central region of human EIF2B1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1967
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.