Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF2b beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100694
Description
Antibody detects endogenous levels of total EIF2B2.
eIF2b beta is the beta subunit of eukaryotic initiation factor-2B (EIF2B). EIF2B is involved in protein synthesis and exchanges GDP and GTP for its activation and deactivation.
Specifications
eIF2b beta | |
Polyclonal | |
Unconjugated | |
Eif2b2 | |
AA409345; C85417; EIF2B; eIF-2B GDP-GTP exchange factor subunit beta; Eif2b2; EIF2BB; EIF-2Bbeta; eukaryotic translation initiation factor 2B subunit beta; eukaryotic translation initiation factor 2B, subunit 2 (beta, 39kD); eukaryotic translation initiation factor 2B, subunit 2 beta; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; translation initiation factor eIF-2B subunit beta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
217715, 84005, 8892 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49770, Q62818, Q99LD9 | |
Eif2b2 | |
A synthesized peptide derived from human EIF2B2(Accession P49770), corresponding to amino acid residues E75-H125. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction