Learn More
Invitrogen™ eIF2b beta Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (94%), Zebrafish (85%), Rabbit (98%), Chicken (90%), Rhesus Monkey (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | eIF2b beta |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Eif2b2 |
| Gene Accession No. | P49770 |
| Gene Alias | AA409345; C85417; EIF2B; eIF-2B GDP-GTP exchange factor subunit beta; Eif2b2; EIF2BB; EIF-2Bbeta; eukaryotic translation initiation factor 2B subunit beta; eukaryotic translation initiation factor 2B, subunit 2 (beta, 39kD); eukaryotic translation initiation factor 2B, subunit 2 beta; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; translation initiation factor eIF-2B subunit beta |
| Gene Symbols | Eif2b2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.