Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF2b beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552217 Supplier Invitrogen™ Supplier No. PA552217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFCQGHEMAV NLSKAGIETT VMTDAAIFAV MSRVNKVIIG TKTILANGAL RAVTGTHTLA LAAKHHSTPL IVCAPMFKLS PQFPNEEDSF HKFVAPEEVL PFTEGDILEK VSVHCPVFDY VPPELITLFI SNIGGNAPSY IYRLMSELYH Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

eIF2b beta is the beta subunit of eukaryotic initiation factor-2B (EIF2B). EIF2B is involved in protein synthesis and exchanges GDP and GTP for its activation and deactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2b beta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif2b2
Gene Accession No. P49770, Q62818, Q99LD9
Gene Alias AA409345; C85417; EIF2B; eIF-2B GDP-GTP exchange factor subunit beta; Eif2b2; EIF2BB; EIF-2Bbeta; eukaryotic translation initiation factor 2B subunit beta; eukaryotic translation initiation factor 2B, subunit 2 (beta, 39kD); eukaryotic translation initiation factor 2B, subunit 2 beta; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; translation initiation factor eIF-2B subunit beta
Gene Symbols Eif2b2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human eIF2b beta. Recombinant protein control fragment (Product #RP-88928).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217715, 84005, 8892
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.