Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF2B4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530292 Supplier Invitrogen™ Supplier No. PA530292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Chimpanzee (97%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF2B4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene EIF2B4
Gene Accession No. Q61749, Q63186, Q9UI10
Gene Alias DKFZp586J0119; EIF2B; EIF-2B; eIF-2B GDP-GTP exchange factor subunit delta; EIF2B4; Eif2bd; EIF2Bdelta; eukaryotic translation initiation factor 2B subunit 4 delta; eukaryotic translation initiation factor 2B subunit delta; eukaryotic translation initiation factor 2B, subunit 4 delta; eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa; Jgr1a; translation initiation factor eIF-2b delta subunit; translation initiation factor eIF-2B subunit delta
Gene Symbols EIF2B4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EIF2B delta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117019, 13667, 8890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.