Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF2B4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565254 Supplier Invitrogen™ Supplier No. PA565254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGREMTKEEKL QLRKEKKQQK KKRKEEKGAE PETGSAVSAA QCQVGPTREL PESGIQLGTP REKVPAGRSK AELRAER Highest antigen sequence identity to the following orthologs - mouse 78%, rat 77%.

Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF2B4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF2B4
Gene Accession No. Q9UI10
Gene Alias DKFZp586J0119; EIF2B; EIF-2B; eIF-2B GDP-GTP exchange factor subunit delta; EIF2B4; Eif2bd; EIF2Bdelta; eukaryotic translation initiation factor 2B subunit 4 delta; eukaryotic translation initiation factor 2B subunit delta; eukaryotic translation initiation factor 2B, subunit 4 delta; eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa; Jgr1a; translation initiation factor eIF-2b delta subunit; translation initiation factor eIF-2B subunit delta
Gene Symbols EIF2B4
Host Species Rabbit
Immunogen Recombinant Human eIF2b delta. Recombinant protein control fragment (Product #RP-104712).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.