Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF2b epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584639 Supplier Invitrogen™ Supplier No. PA584639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SILEENVLLG SGTVIGSNCF ITNSVIGPGC HIGDNVVLDQ TYLWQGVRVA AGAQIHQSLL CDNAEVKERV TLKPRSVLTS QVVVGPNITL PEGSVISLHP PD Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 88%.

This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2b epsilon
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif2b5
Gene Accession No. Q13144
Gene Alias C81315; CACH; CLE; EIF-2B; eIF-2B GDP-GTP exchange factor subunit epsilon; EIF2B5; EIF2BE; EIF2Bepsilon; eukaryotic translation initiation factor 2B subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; LVWM; Translation initiation factor eIF-2B subunit epsilon
Gene Symbols Eif2b5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EIF2B5. Recombinant protein control fragment (Product #RP-107331).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.