Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF2b epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120078 Supplier Invitrogen™ Supplier No. PA5120078
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, A-549, HL-60, HT-1080, LO2, Mouse brain. Immunogen sequence: TLPEGSVISL HPPDAEEDED DGEFSDDSGA DQEKDKVKMK GYNPAEVGAA GKGYLWKAAG MNMEEEEELQ QNLWGLKINM EEESESESEQ SMDSEEPDSR GGSPQMDDIK VFQNEVLGTL QRGKEENISC DNLVLEINSL KYAYNISLKE VMQVLSHVVL EFPLQQMDSP LDSSRYCALL LPLLKAWSPV FRNYIKRAAD HLEALAAIED FFLEHEALGI SMAKVLMAFY QLEILAEETI LSWFSQRDTT DKGQQLRKNQ QLQRFIQWLK EAEEESSEDD.

This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2b epsilon
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Eif2b5
Gene Accession No. Q13144, Q8CHW4
Gene Alias C81315; CACH; CLE; EIF-2B; eIF-2B GDP-GTP exchange factor subunit epsilon; EIF2B5; EIF2BE; EIF2Bepsilon; eukaryotic translation initiation factor 2B subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; LVWM; Translation initiation factor eIF-2B subunit epsilon
Gene Symbols Eif2b5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 442-721 of human eIF2B epsilon (eIF2B epsilon (EIF2B5)) (NP_0038982).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224045, 8893
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.