Learn More
Invitrogen™ eIF2b gamma Polyclonal Antibody

Description
This antibody is predicted to react with non-human primate based on sequence homology.
Specifications
Specifications
| Antigen | eIF2b gamma |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EIF2B3 |
| Gene Accession No. | Q9NR50 |
| Gene Alias | 1190002P15Rik; EIF-2B; eIF-2B GDP-GTP exchange factor subunit gamma; EIF2B3; Eif2bg; EIF2Bgamma; eukaryotic translation initiation factor 2B subunit gamma; eukaryotic translation initiation factor 2B, subunit 3; eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD); eukaryotic translation initiation factor 2B, subunit 3 gamma; eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa; translation initiation factor eIF-2B subunit gamma; Translation initiation factor eIF-2B subunit gamma-like protein |
| Gene Symbols | EIF2B3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.