Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF2B1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093780 20 μL
NB093779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093780 Supplier Novus Biologicals Supplier No. NBP30465920UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF2B1 Polyclonal specifically detects eIF2B1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen eIF2B1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EIF2B, EIF-2B, eIF-2B GDP-GTP exchange factor subunit alpha, EIF-2Balpha, EIF2BAMGC117409, eukaryotic translation initiation factor 2B, subunit 1 (alpha, 26kD), eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa, MGC125868, MGC125869, translation initiation factor eIF-2B subunit alpha
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-305 of human EIF2B1 (NP_001405.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1967
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.