Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF2B3 Antibody (OTI1A4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271397H
Description
eIF2B3 Monoclonal specifically detects eIF2B3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
eIF2B3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
EIF2B3 | |
Full length recombinant protein of human eIF2B3 (NP_065098) produced in HEK293T cells. | |
0.1 mL | |
8891.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1A4 | |
Western Blot, Immunohistochemistry | |
EIF-2B, eIF-2B GDP-GTP exchange factor subunit gamma, EIF2Bgamma, eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD), eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa, translation initiation factor eIF-2B subunit gamma | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction