Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2B3 Monoclonal Antibody (OTI1H3), TrueMAB™, OriGene
SDP

Catalog No. 502540839
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0839 30 μL
50-167-4423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0839 Supplier OriGene Supplier No. TA503562S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EIF2B3 Monoclonal antibody specifically detects EIF2B3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen EIF2B3
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H3
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SEC14L2
Gene Accession No. O76054
Gene Alias 1190002P15Rik; EIF-2B; eIF-2B GDP-GTP exchange factor subunit gamma; EIF2B3; Eif2bg; EIF2Bgamma; eukaryotic translation initiation factor 2B subunit gamma; eukaryotic translation initiation factor 2B, subunit 3; eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD); eukaryotic translation initiation factor 2B, subunit 3 gamma; eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa; translation initiation factor eIF-2B subunit gamma; Translation initiation factor eIF-2B subunit gamma-like protein
Host Species Mouse
Immunogen Full length human recombinant protein of human SEC14L2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23541
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.