Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2B3 Mouse anti-Human, Rat, Clone: 1H3, Abnova™

Catalog No. 89035445 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-445 Supplier Abnova Corporation Supplier No. MAB14040

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human EIF2B3.

  • High specificity
  • High reactivity

Specifications

Antigen EIF2B3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human EIF2B3.
Dilution Immunocytochemistry (1:10-1:100) Immunofluorescence (1:10-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) Immunoprecipitation (3 ug) Western Blot (1:1000) The optimal working dilution should be dete
Formulation In PBS (30% glycerol, 1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. Q9NR50
Gene Alias EIF-2B/EIF2Bgamma
Gene Symbols EIF2B3
Host Species Mouse
Immunogen Full length human eIF2B3 produced in HEK293T cells.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8891
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.