Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EIF2B4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144963
Description
Immunogen sequence: VGREMTKEEK LQLRKEKKQQ KKKRKEEKGA EPETGSAVSA AQCQVGPTRE LPESGIQLGT PREKVPAGRS KAELRAER Antigen sequence identity: Mouse 78%; Rat 77%.
Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
EIF2B4 | |
Polyclonal | |
Unconjugated | |
EIF2B4 | |
DKFZp586J0119; EIF2B; EIF-2B; eIF-2B GDP-GTP exchange factor subunit delta; EIF2B4; Eif2bd; EIF2Bdelta; eukaryotic translation initiation factor 2B subunit 4 delta; eukaryotic translation initiation factor 2B subunit delta; eukaryotic translation initiation factor 2B, subunit 4 delta; eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa; Jgr1a; translation initiation factor eIF-2b delta subunit; translation initiation factor eIF-2B subunit delta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8890 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UI10 | |
EIF2B4 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 27-104. | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction