Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF2C3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 196921AP150UL
Description
This antibody is specific to EIF2C3.
Small interfering RNAs (siRNAs) have gained much attention for their powerful ability to suppress gene expression. Introduction of double-stranded RNA (dsRNA), that are homologues in sequence to a gene, has proven to suppress that gene′s expression through a process known as RNA interference (RNAi); this process post-transcriptionally silences a gene through mRNA inhibition or degradation. Both biochemical and genetic studies have led to the identification of two, conserved gene families that are universal components of the interference process. These are the Dicer family, which is comprised of members of the Ribonuclease III family of enzymes, and the Argonaute gene family. Argonaute proteins addition to their role in RNAi process, may also influence development, such as, stem cell fate determination.Specifications
| EIF2C3 | |
| Polyclonal | |
| Unconjugated | |
| AGO3 | |
| AGO3, Argonaute 3, Argonaute3, eIF 2C 3, eIF2C 3, EIF2C3, hAgo3, Protein argonaute 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192669, 214150, 313593 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CJF9, Q9H9G7 | |
| Ago3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction