Learn More
Invitrogen™ eIF2d Polyclonal Antibody

Description
eIF2d Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | eIF2d |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | EIF2D |
| Gene Accession No. | Q61211 |
| Gene Alias | D1Ertd5e; Eif2d; Eukaryotic translation initiation factor 2D; HCA56; Hepatocellular carcinoma-associated antigen 56; LGTN; Ligatin |
| Gene Symbols | EIF2D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.