Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF2d Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555799
Description
Immunogen sequence: HQAREDKSLS EAPEDTSTRG LNQDSTDSKT LQEQMDELLQ QCFLHALKCR VKKADLPLLT STFLGSHMFS CCPEGRQLDI KKSSYKKLSK FLQQMQQEQ Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 75%.
This gene encodes a translation initiation factor involved in the recruitment and delivery of aminoacyl-tRNAs to the P-site of the eukaryotic ribosome in a GTP-independent manner. This gene was previously referred to as ligatin, but is now known to localize to the cytoplasm and localize and function with translation factors. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
eIF2d | |
Polyclonal | |
Unconjugated | |
EIF2D | |
D1Ertd5e; Eif2d; Eukaryotic translation initiation factor 2D; HCA56; Hepatocellular carcinoma-associated antigen 56; LGTN; Ligatin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P41214 | |
EIF2D | |
Recombinant protein corresponding to Human eIF2d. Recombinant protein control fragment (Product #RP-94291). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction