Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF2S1 Monoclonal Antibody (OTI1H1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501402S


Description
EIF2S1 Monoclonal antibody specifically detects EIF2S1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
EIF2S1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P05198 | |
Mouse | |
Affinity chromatography | |
RUO | |
1965 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1H1 | |
Unconjugated | |
EIF2S1 | |
0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
Full length human recombinant protein of human EIF2S1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction