Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2S1 Monoclonal Antibody (OTI5A5), TrueMAB™, OriGene
SDP

Catalog No. 502542337
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2337 30 μL
50-167-2517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2337 Supplier OriGene Supplier No. TA501374S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EIF2S1 Monoclonal antibody specifically detects EIF2S1 in Canine, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen EIF2S1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5A5
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene EIF2S1
Gene Accession No. P05198, P68101
Gene Alias 0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01
Host Species Mouse
Immunogen Full length human recombinant protein of human EIF2S1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1965, 480361, 54318
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.