Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $256.00
Specifications
Antigen | EIF3A |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.25-1.0 ug/ml, Simple Western 1:50, Immunohistochemistry-Paraffin 5-10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224570SS
![]() |
Novus Biologicals
NBP224570SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224570
![]() |
Novus Biologicals
NBP224570 |
0.1 mg |
Each for $256.00
|
|
|||||
Description
EIF3A Polyclonal specifically detects EIF3A in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EIF3A | |
Western Blot 0.25-1.0 ug/ml, Simple Western 1:50, Immunohistochemistry-Paraffin 5-10 ug/ml | |
Polyclonal | |
Rabbit | |
Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
centrosomin homolog, cytoplasmic protein p167, eIF3 p167, eIF3 p180, eIF3 p185, EIF3, p180 subunit, eIF3a, eIF3-p170, EIF3S10EIF3, eIF-3-theta, eIF3-theta, Eukaryotic translation initiation factor 3 subunit 10, eukaryotic translation initiation factor 3 subunit A, eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD), eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD), eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa, eukaryotic translation initiation factor 3, subunit 10, 170kD, eukaryotic translation initiation factor 3, subunit A, KIAA0139P167, p180, p185, TIF32 | |
EIF3A | |
IgG | |
Affinity Purified | |
Reacts in human and mouse. Predicted to work in other species based on homology. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
Q14152 | |
8661 | |
A portion of amino acids 550-600 of human eIF3A was used as the immunogen for the antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title