Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3A Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735731 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5731 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5731 Supplier Proteintech Group Inc Supplier No. 276651AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The German cDNA consortium.

Specifications

Antigen EIF3A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EIF3A
Gene Accession No. Q14152
Gene Alias eIF 3 theta, EIF3, eIF3 p167, eIF3 p170, eIF3 p180, eIF3 p185, eIF3 theta, EIF3A, EIF3S10, KIAA0139, P167, p180, p185, TIF32
Gene Symbols EIF3A
Host Species Rabbit
Immunogen EIF3A Fusion Protein Ag24752
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8661
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.