Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3B/EIF3S9 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572488
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2488 10 μL
50-157-2487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2488 Supplier Bethyl Laboratories, Inc Supplier No. IHC00659T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

EIF3S9 binds to the 40S ribosome and promotes the binding of methionyl-tRNAi and mRNA.

Specifications

Antigen eIF3B/EIF3S9
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EIF3B
Gene Accession No. P55884, Q8JZQ9
Gene Alias EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, MGC104664, MGC131875, PRT1, eIF-3-eta; eIF3 p110; eIF3 p116; eukaryotic translation initiation factor 3 subunit 9; eukaryotic translation initiation factor 3 subunit B; eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD); eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa; hPrt1; prt1 homolog
Gene Symbols Eif3b
Host Species Rabbit
Immunogen Between 100 and 150
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27979, 8662
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.