Learn More
eIF3B/EIF3S9 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301760A

Description
The recommended shelf life for this product is 1 year from date of receipt.
EIF3S9 binds to the 40S ribosome and promotes the binding of methionyl-tRNAi and mRNA.Specifications
eIF3B/EIF3S9 | |
Polyclonal | |
Unconjugated | |
EIF3B | |
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, MGC104664, MGC131875, PRT1, eIF-3-eta; eIF3 p110; eIF3 p116; eukaryotic translation initiation factor 3 subunit 9; eukaryotic translation initiation factor 3 subunit B; eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD); eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa; hPrt1; prt1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27979, 8662 | |
4° C | |
Liquid |
Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P55884, Q8JZQ9 | |
EIF3B | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.