Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF3C Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100511JF646
Description
EIF3C Polyclonal antibody specifically detects EIF3C in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
EIF3C | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
cell migration-inducing protein 17, eIF3 p110, eIF3c, EIF3CL, eIF3-p110, EIF3S8FLJ55450, Eukaryotic translation initiation factor 3 subunit 8, eukaryotic translation initiation factor 3 subunit C, eukaryotic translation initiation factor 3, subunit 8 (110kD), eukaryotic translation initiation factor 3, subunit 8, 110kDa, eukaryotic translation initiation factor 3, subunit C, FLJ53378, FLJ54400, FLJ54404, FLJ55750, FLJ78287, MGC189737, MGC189744 | |
A synthetic peptide derived from a sequence between residues 850 and 918 (the C-terminus) of human eukaryotic translation initiation factor 3, subunit 8, 110kDa using the numbering given in Swiss-Prot entry Q99613 (GeneID 8663). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8663 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction