Learn More
Invitrogen™ eIF3c Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (93%), Xenopus laevis (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | eIF3c |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Eif3c |
| Gene Accession No. | Q99613 |
| Gene Alias | 110kDa; 3230401O13Rik; cell migration-inducing protein 17; eIF3 p110; eIF3 p110 {ECO:0000255; eIF3c; eIF3c {ECO:0000255; EIF3CL; eIF3-p110; EIF3S8; eukaryotic translation initiation factor 3 subunit 8; eukaryotic translation initiation factor 3 subunit 8 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit C; eukaryotic translation initiation factor 3 subunit C {ECO:0000255; eukaryotic translation initiation factor 3, subunit 8 (110 kDa); eukaryotic translation initiation factor 3, subunit 8 (110kD); eukaryotic translation initiation factor 3, subunit 8, 110kDa; eukaryotic translation initiation factor 3, subunit C; extra-toes spotting; HAMAP-Rule:MF_03002}; NIPIL(A3); NipilA3; NIPI-like protein; Xs; Xsl |
| Gene Symbols | Eif3c |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.