Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3c Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561425
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561425 Supplier Invitrogen™ Supplier No. PA561425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YYKFDYKAHQ RQLTPPEGSS KSEQDQAENE GEDSAVLMER LCKYIYAKDR TDRIRTCAIL CHIYHHALHS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3c
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif3c
Gene Accession No. Q99613
Gene Alias 110kDa; 3230401O13Rik; cell migration-inducing protein 17; eIF3 p110; eIF3 p110 {ECO:0000255; eIF3c; eIF3c {ECO:0000255; EIF3CL; eIF3-p110; EIF3S8; eukaryotic translation initiation factor 3 subunit 8; eukaryotic translation initiation factor 3 subunit 8 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit C; eukaryotic translation initiation factor 3 subunit C {ECO:0000255; eukaryotic translation initiation factor 3, subunit 8 (110 kDa); eukaryotic translation initiation factor 3, subunit 8 (110kD); eukaryotic translation initiation factor 3, subunit 8, 110kDa; eukaryotic translation initiation factor 3, subunit C; extra-toes spotting; HAMAP-Rule:MF_03002}; NIPIL(A3); NipilA3; NIPI-like protein; Xs; Xsl
Gene Symbols Eif3c
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human eIF3c. Recombinant protein control fragment (Product #RP-104421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8663
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.