Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF3D Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297518AF350
Description
EIF3D Polyclonal specifically detects EIF3D in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
EIF3D | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8664 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 350 | |
eIF3d, eIF3-p66, EIF3S7eIF3 p66, eIF-3-zeta, eIF3-zeta, Eukaryotic translation initiation factor 3 subunit 7, eukaryotic translation initiation factor 3 subunit D, eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa, eukaryotic translation initiation factor 3, subunit D, MGC126526, MGC17258, translation initiation factor eIF3 p66 subunit | |
Produced in rabbits immunized with E. coli-derived Human EIF3D fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction