Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF3d Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101587
Description
Antibody detects endogenous levels of total EIF3D.
eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.
Specifications
eIF3d | |
Polyclonal | |
Unconjugated | |
EIF3D | |
66/67kDa; AA407891; actinin alpha 4; ACTININ-4; Actn4; alpha actinin 4; alpha-actinin-4; eIF3 p66; EIF3D; eIF3d {ECO:0000255; eIF3p66; eIF3-p66; Eif3s7; eIF-3-zeta; eIF3-zeta; eIF-3-zeta {ECO:0000255; eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta; eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; HAMAP-Rule:MF_03003}; non-muscle alpha-actinin 4; RP5-1119A7.12-003; si:dkey-165I8.6; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit; wu:fb12a03; zgc:64097 | |
Rabbit | |
Affinity chromatography | |
RUO | |
362952, 55944, 8664 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15371, O70194, Q6AYK8 | |
EIF3D | |
A synthesized peptide derived from human EIF3D(Accession O15371), corresponding to amino acid residues Q102-Q152. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction