Learn More
Invitrogen™ eIF3e Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595431
Description
Human EIF3e shares 100% amino acid (aa) sequence identity with both mouse and rat EIF3e. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat thymus tissue, mouse thymus tissue, mouse spleen tissue, COLO320 whole cell, HELA whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. Required for nonsense-mediated mRNA decay (NMD); may act in conjunction with UPF2 to divert mRNAs from translation to the NMD pathway. May interact with MCM7 and EPAS1 and regulate the proteasome-mediated degradation of these proteins.
Specifications
eIF3e | |
Polyclonal | |
Unconjugated | |
EIF3E | |
48kDa; eIF-3 p48; eIF3e; eIF3-p46; EIF3-P48; EIF3S6; eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3 subunit E-like protein; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; eukaryotic translation initiation factor subunit E-like protein; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; viral integration site protein INT-6 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
103692906, 16341, 3646 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P60228, P60229, Q641X8 | |
EIF3E | |
E.coli-derived human EIF3e recombinant protein (Position: A160-Q241). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.