Learn More
Invitrogen™ eIF3e Recombinant Rabbit Monoclonal Antibody (3H4N2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538067
Description
Positive Samples: HeLa, 293T, Jurkat, Mouse lung, Mouse testis, Rat lung, Rat testis Immunogen sequence: QRTWLIHWSL FVFFNHPKGR DNIIDLFLYQ PQYLNAIQTM CPHILRYLTT AVITNKDVRK RRQVLKDLVK VIQQESYTYK DPITEFVECL YVNFDFDGAQ KKLRECESVL VNDFFLVACL EDFIENARLF IFETFCRIHQ C.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. Required for nonsense-mediated mRNA decay (NMD); may act in conjunction with UPF2 to divert mRNAs from translation to the NMD pathway. May interact with MCM7 and EPAS1 and regulate the proteasome-mediated degradation of these proteins.
Specifications
eIF3e | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60228, P60229, Q641X8 | |
EIF3E | |
Recombinant fusion protein containing a sequence corresponding to amino acids 210-350 of human eIF3e (P60228). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3H4N2 | |
Unconjugated | |
EIF3E | |
48kDa; eIF-3 p48; eIF3e; eIF3-p46; EIF3-P48; EIF3S6; eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3 subunit E-like protein; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; eukaryotic translation initiation factor subunit E-like protein; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; viral integration site protein INT-6 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
103692906, 16341, 3646 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.