Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3e Recombinant Rabbit Monoclonal Antibody (JE53-63)
GREENER_CHOICE

Catalog No. PIMA536163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536163 Supplier Invitrogen™ Supplier No. MA536163
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

eIF3e Recombinant Monoclonal Antibody for IHC (P), Western Blot

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. Required for nonsense-mediated mRNA decay (NMD); may act in conjunction with UPF2 to divert mRNAs from translation to the NMD pathway. May interact with MCM7 and EPAS1 and regulate the proteasome-mediated degradation of these proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3e
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE53-63
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene EIF3E
Gene Accession No. P60228, P60229, Q641X8
Gene Alias 48kDa; eIF-3 p48; eIF3e; eIF3-p46; EIF3-P48; EIF3S6; eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3 subunit E-like protein; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; eukaryotic translation initiation factor subunit E-like protein; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; viral integration site protein INT-6 homolog
Gene Symbols EIF3E
Host Species Rabbit
Immunogen Recombinant protein within Human eIF3e aa 190-370.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103692906, 16341, 3646
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.