Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3f Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596549 Supplier Invitrogen™ Supplier No. PA596549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGRVVRLHPV ILASIVDSYE RRNEGAARVI GTLLGTVDKH SVEVTNCFSV PHNESEDEVA VDMEFAKNMY ELHKKVSPNE LILGWYATGH DITEHSVLIH EYYSREAPNP IHLTVDTSLQ NGRMSIKAYV STLMGVPGRT MGVMFTPLTV KYAYYDTERI GVDLIMKTCF SPNRVIGLSS DLQQVGGASA RIQDALSTVL QYAEDVLSGK VSADNTVGRF LMSLVNQVPK IVPDDFETML NSNINDLLMV TYLANLTQSQ IALNEKLVNL; Positive Samples: HL-60, SKOV3, BT-474, Jurkat, HeLa, Mouse spleen, Mouse lung, Mouse heart, Rat brain; Cellular Location: Cytoplasm.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3f
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EIF3F
Gene Accession No. O00303, Q9DCH4
Gene Alias 0610037M02Rik; AA409853; deubiquitinating enzyme eIF3f; eIF3 p47; eIF-3-epsilon; eIF3-epsilon; EIF3F; eIF3-p47; Eif3s5; eukaryotic translation initiation factor 3 subunit 5; eukaryotic translation initiation factor 3 subunit 5 epsilon; eukaryotic translation initiation factor 3 subunit F; eukaryotic translation initiation factor 3, subunit 5 (epsilon); eukaryotic translation initiation factor 3, subunit 5 (epsilon, 47kD); eukaryotic translation initiation factor 3, subunit 5 epsilon, 47kDa; eukaryotic translation initiation factor 3, subunit F
Gene Symbols EIF3F
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 88-357 of human EIF3F (NP_0037451).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293427, 66085, 8665
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.