Learn More
Invitrogen™ eIF3f Polyclonal Antibody

Description
The antibody detects endogenous levels of total EIF3F protein.
Specifications
Specifications
| Antigen | eIF3f |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | EIF3F |
| Gene Accession No. | O00303, Q9DCH4 |
| Gene Alias | 0610037M02Rik; AA409853; deubiquitinating enzyme eIF3f; eIF3 p47; eIF-3-epsilon; eIF3-epsilon; EIF3F; eIF3-p47; Eif3s5; eukaryotic translation initiation factor 3 subunit 5; eukaryotic translation initiation factor 3 subunit 5 epsilon; eukaryotic translation initiation factor 3 subunit F; eukaryotic translation initiation factor 3, subunit 5 (epsilon); eukaryotic translation initiation factor 3, subunit 5 (epsilon, 47kD); eukaryotic translation initiation factor 3, subunit 5 epsilon, 47kDa; eukaryotic translation initiation factor 3, subunit F |
| Gene Symbols | EIF3F |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.