Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3g Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541774 Supplier Invitrogen™ Supplier No. PA541774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRDGASRRGE SMQPNRRADD NATIRVTNLS EDTRETDLQE LFRPFGSISR Sequence homology: Cow: 100%; Dog: 100%; Goat: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of posttermination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. This subunit can bind 18S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3g
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EIF3G
Gene Accession No. O75821, Q9Z1D1
Gene Alias 44kDa; D0Jmb4; eiF3 delta; eIF3 p42; eIF3 p42 {ECO:0000255; eIF3 p44; eIF3 p44 {ECO:0000255; eIF-3 RNA-binding subunit; eIF-3 RNA-binding subunit {ECO:0000255; eIF-3-delta; eIF3-delta; eIF-3-delta {ECO:0000255; EIF3G; eIF3g {ECO:0000255; Eif3p42; EIF3-P42; eIF3-p44; EIF3S4; eukaryotic translation initiation factor 3 RNA-binding subunit; eukaryotic translation initiation factor 3 RNA-binding subunit {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 4; eukaryotic translation initiation factor 3 subunit 4 {ECO:0000255; eukaryotic translation initiation factor 3 subunit G; eukaryotic translation initiation factor 3 subunit G {ECO:0000255; eukaryotic translation initiation factor 3, subunit 4 (delta); eukaryotic translation initiation factor 3, subunit 4 (delta, 44 kDa); eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD); eukaryotic translation initiation factor 3 subunit p42; eukaryotic translation initiation factor 3, subunit 4 delta; eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa; eukaryotic translation initiation factor 3, subunit G; HAMAP-Rule:MF_03006}; p44; similar to eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa; translation initiation factor eIF3-p44; TU-189B2; Unknown (protein for MGC:128341); zgc:56553
Gene Symbols EIF3G
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human EIF3G.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53356, 8666
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.