Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF3G Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299199R
Description
EIF3G Polyclonal specifically detects EIF3G in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
EIF3G | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
8666 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
eIF3 p42, eIF3 p44, eIF-3-delta, eIF3-delta, eIF3g, EIF3-P42, eIF3-p44, EIF3S4eIF-3 RNA-binding subunit, Eukaryotic translation initiation factor 3 RNA-binding subunit, Eukaryotic translation initiation factor 3 subunit 4, eukaryotic translation initiation factor 3 subunit G, eukaryotic translation initiation factor 3 subunit p42, eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD), eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa, eukaryotic translation initiation factor 3, subunit G | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human EIF3G. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction