Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF3h Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528516
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (97%), Chicken (91%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
Specifications
| eIF3h | |
| Polyclonal | |
| Unconjugated | |
| Eif3h | |
| 1110008A16Rik; 40 kDa; 40kD; 9430017H16Rik; eIF-3 gamma; eIF3 p40 subunit; eIF3 p40 subunit {ECO:0000255; eIF-3-gamma; eIF3-gamma; EIF3H; eIF3h {ECO:0000255; eIF3p40; eIF3-p40; EIF3S3; eukaryotic translation initiation factor 3 subunit 3; eukaryotic translation initiation factor 3 subunit 3 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 3 gamma; eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3 subunit H {ECO:0000255; eukaryotic translation initiation factor 3 subunit H-like protein; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; eukaryotic translation initiation factor subunit H-like protein; HAMAP-Rule:MF_03007}; MGC102958 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.87 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O15372 | |
| Eif3h | |
| Recombinant protein encompassing a sequence within the center region of human eIF3H/EIF3S3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction