Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3h Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543574 Supplier Invitrogen™ Supplier No. PA543574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MASRKEGTGS TATSSSSTAG AAGKGKGKGG SGDSAVKQVQ IDGLVVLKII Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%.

eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3h
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Eif3h
Gene Accession No. O15372, Q91WK2
Gene Alias 1110008A16Rik; 40 kDa; 40kD; 9430017H16Rik; eIF-3 gamma; eIF3 p40 subunit; eIF3 p40 subunit {ECO:0000255; eIF-3-gamma; eIF3-gamma; EIF3H; eIF3h {ECO:0000255; eIF3p40; eIF3-p40; EIF3S3; eukaryotic translation initiation factor 3 subunit 3; eukaryotic translation initiation factor 3 subunit 3 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 3 gamma; eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3 subunit H {ECO:0000255; eukaryotic translation initiation factor 3 subunit H-like protein; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; eukaryotic translation initiation factor subunit H-like protein; HAMAP-Rule:MF_03007}; MGC102958
Gene Symbols Eif3h
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human EIF3H (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68135, 8667
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.