Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF3h Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587290
Description
Purity is > 95% by SDS-PAGE.
eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
Specifications
eIF3h | |
Polyclonal | |
Unconjugated | |
Eif3h | |
1110008A16Rik; 40 kDa; 40kD; 9430017H16Rik; eIF-3 gamma; eIF3 p40 subunit; eIF3 p40 subunit {ECO:0000255; eIF-3-gamma; eIF3-gamma; EIF3H; eIF3h {ECO:0000255; eIF3p40; eIF3-p40; EIF3S3; eukaryotic translation initiation factor 3 subunit 3; eukaryotic translation initiation factor 3 subunit 3 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 3 gamma; eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3 subunit H {ECO:0000255; eukaryotic translation initiation factor 3 subunit H-like protein; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; eukaryotic translation initiation factor subunit H-like protein; HAMAP-Rule:MF_03007}; MGC102958 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
299899, 68135, 8667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O15372, Q6P9U8, Q91WK2 | |
Eif3h | |
Recombinant full length Human EIF3H. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction