Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3i Monoclonal Antibody (3B10)
GREENER_CHOICE

Catalog No. PIMA520471
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520471 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520471 Supplier Invitrogen™ Supplier No. MA520471
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: FRTERPVNSA ALSPNYDHVV LGGGQEAMDV TTTSTRIGKF EARFFHLAFE EEFGRVKGHF GPINSVAFHP DGKSYSSGGE DGYVRIHYFD PQYFEFEFEA.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3i
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Eif3i
Gene Accession No. Q13347
Gene Alias 36kDa; D4Ertd632e; eIF3 p36; eIF-3-beta; eIF3-beta; Eif3i; eIF3-p36; Eif3s2; Eukaryotic translation initiation factor 3 subunit 2; eukaryotic translation initiation factor 3 subunit I; eukaryotic translation initiation factor 3, subunit 1; eukaryotic translation initiation factor 3, subunit 2 (beta); eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 2 beta, 36kDa; eukaryotic translation initiation factor 3, subunit I; predicted protein of HQ2242; PRO2242; subuni; TGF-beta receptor binding protein; TGF-beta receptor-interacting protein 1; TGFbeta receptor-interacting protein 1; TRIP1; TRIP-1
Gene Symbols Eif3i
Host Species Mouse
Immunogen EIF3I (NP_003748.1, 226 a.a. ~ 325 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8668
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.