Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3i Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556302 Supplier Invitrogen™ Supplier No. PA556302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KYNREGDLLF TVAKDPIVNV WYSVNGERLG TYMGHTGAVW CVDADWDTKH VLTGSADNSC RLWDCETGKQ LALLKTNSAV RTCG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 87%.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3i
Applications Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif3i
Gene Accession No. B0BNA7, Q13347, Q9QZD9
Gene Alias 36kDa; D4Ertd632e; eIF3 p36; eIF-3-beta; eIF3-beta; Eif3i; eIF3-p36; Eif3s2; Eukaryotic translation initiation factor 3 subunit 2; eukaryotic translation initiation factor 3 subunit I; eukaryotic translation initiation factor 3, subunit 1; eukaryotic translation initiation factor 3, subunit 2 (beta); eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 2 beta, 36kDa; eukaryotic translation initiation factor 3, subunit I; predicted protein of HQ2242; PRO2242; subuni; TGF-beta receptor binding protein; TGF-beta receptor-interacting protein 1; TGFbeta receptor-interacting protein 1; TRIP1; TRIP-1
Gene Symbols Eif3i
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human eIF3i. Recombinant protein control fragment (Product #RP-94035).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54709, 682390, 8668
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.